Assay Design



Gene Transcripts

Gene Symbol
Slc25a29
Entrez Gene ID
314441
Gene Name
solute carrier family 25 (mitochondrial carnitine/acylcarnitine carrier), member 29
Gene Aliases
-
Chromosome Location
Chr.6: 132736968 - 132747762 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010958.1 NP_001010958.1 1 - 2 110 60
XM_006240541.2 XP_006240603.1 1 - 2 57 60
GenBank mRNA BC089065.1 - 1 - 2 110 60

View Full Product Details