Assay Design



Gene Transcripts

Gene Symbol
Slc25a29
Entrez Gene ID
314441
Gene Name
solute carrier family 25 (mitochondrial carnitine/acylcarnitine carrier), member 29
Gene Aliases
-
Chromosome Location
Chr.6: 132736968 - 132747762 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010958.1 NP_001010958.1 2 - 3 153 63
GenBank mRNA BC089065.1 - 2 - 3 153 63

View Full Product Details