Assay Design



Gene Transcripts

Gene Symbol
Stra8
Entrez Gene ID
500079
Gene Name
stimulated by retinoic acid 8
Gene Aliases
RGD1562852
Chromosome Location
Chr.4: 62447724 - 62474780 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006236282.2 XP_006236344.1 3 - 4 2709 65
XM_575429.4 XP_575429.2 2 - 3 452 65

View Full Product Details