Assay Design



Gene Transcripts

Gene Symbol
Efna2
Entrez Gene ID
84358
Gene Name
ephrin A2
Gene Aliases
-
Chromosome Location
Chr.7: 12382636 - 12393266 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168670.1 NP_001162141.1 3 - 4 746 83

View Full Product Details