Assay Design



Gene Transcripts

Gene Symbol
Klra1
Entrez Gene ID
494194
Gene Name
killer cell lectin-like receptor, subfamily A, member 1
Gene Aliases
Klra2, Ly49i8
Chromosome Location
Chr.4: 165426365 - 165460142 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009486.1 NP_001009486.1 7 - 8 843 85
XM_008763404.1 XP_008761626.1 6 - 7 820 85
GenBank mRNA AY649835.1 - 7 - 8 843 85
BC085917.1 - - 806 85

View Full Product Details