Assay Design



Gene Transcripts

Gene Symbol
Sdr16c6
Entrez Gene ID
502939
Gene Name
short chain dehydrogenase/reductase family 16C, member 6
Gene Aliases
RGD1562060
Chromosome Location
Chr.5: 16972521 - 16996010 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109356.1 NP_001102826.1 2 - 3 415 102

View Full Product Details