Assay Design



Gene Transcripts

Gene Symbol
Larp4b
Entrez Gene ID
307070
Gene Name
La ribonucleoprotein domain family, member 4B
Gene Aliases
Larp5, RGD1310713
Chromosome Location
Chr.17: 63236007 - 63317298 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107361.1 NP_001100831.1 3 - 4 381 79

View Full Product Details