Assay Design



Gene Transcripts

Gene Symbol
Phf13
Entrez Gene ID
313742
Gene Name
PHD finger protein 13
Gene Aliases
-
Chromosome Location
Chr.5: 169161412 - 169167831 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107995.1 NP_001101465.1 3 - 4 970 67

View Full Product Details