Assay Design



Gene Transcripts

Gene Symbol
Zbtb8a
Entrez Gene ID
313049
Gene Name
zinc finger and BTB domain containing 8a
Gene Aliases
Zbtb8
Chromosome Location
Chr.5: 147554907 - 147584281 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107913.1 NP_001101383.1 2 - 3 364 90

View Full Product Details