Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013168.1 | NP_001013186.1 | 5 - 6 | 418 | 69 |
GenBank mRNA | BC081863.1 | - | 5 - 6 | 418 | 69 |
FQ214236.1 | - | 5 - 6 | 420 | 69 | |
FQ231836.1 | - | 5 - 6 | 415 | 69 |