Assay Design
Gene Transcripts
- Gene Symbol
- Pithd1
- Entrez Gene ID
-
298557
- Gene Name
- PITH (C-terminal proteasome-interacting domain of thioredoxin-like) domain containing 1
- Gene Aliases
-
RGD1308048
- Chromosome Location
-
Chr.5: 154322562 - 154333095 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_216543.7 |
XP_216543.5 |
1 - 2 |
323 |
74 |
View Full Product Details