Assay Design



Gene Transcripts

Gene Symbol
Slc46a2
Entrez Gene ID
298034
Gene Name
solute carrier family 46, member 2
Gene Aliases
Tscot
Chromosome Location
Chr.5: 77165492 - 77173761 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106652.1 NP_001100122.1 1 - 2 1269 68

View Full Product Details