Assay Design
Gene Transcripts
- Gene Symbol
- Zscan5b
- Entrez Gene ID
-
292566
- Gene Name
- zinc finger and SCAN domain containing 5B
- Gene Aliases
-
RGD1559143
- Chromosome Location
-
Chr.1: 71334391 - 71342633 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_218239.5 |
XP_218239.5 |
2 - 3 |
566 |
80 |
View Full Product Details