Assay Design



Gene Transcripts

Gene Symbol
Arhgap40
Entrez Gene ID
296323
Gene Name
Rho GTPase activating protein 40
Gene Aliases
RGD1562428
Chromosome Location
Chr.3: 154996720 - 155042028 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008762437.1 XP_008760659.1 13 - 14 1903 67

View Full Product Details