Assay Design



Gene Transcripts

Gene Symbol
Acot8
Entrez Gene ID
170588
Gene Name
acyl-CoA thioesterase 8
Gene Aliases
Pte1
Chromosome Location
Chr.3: 161260837 - 161272495 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130756.3 NP_570112.3 5 - 6 1241 66
GenBank mRNA AF452100.1 - 5 - 6 891 66
BC078751.1 - 5 - 6 1235 66

View Full Product Details