Assay Design



Gene Transcripts

Gene Symbol
Ift52
Entrez Gene ID
362265
Gene Name
intraflagellar transport 52
Gene Aliases
RGD1311004
Chromosome Location
Chr.3: 159388868 - 159413358 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177685.1 NP_001171156.1 1 - 2 52 64
GenBank mRNA BC162007.1 - 1 - 2 52 64

View Full Product Details