Assay Design



Gene Transcripts

Gene Symbol
Rab5c
Entrez Gene ID
287709
Gene Name
RAB5C, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.10: 88621924 - 88645364 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105840.2 NP_001099310.2 4 - 5 635 81
GenBank mRNA BC158856.1 - 4 - 5 617 81

View Full Product Details