Assay Design



Gene Transcripts

Gene Symbol
Afmid
Entrez Gene ID
688283
Gene Name
arylformamidase
Gene Aliases
-
Chromosome Location
Chr.10: 106828965 - 106844314 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111366.1 NP_001104836.1 1 - 2 85 86
XM_006247823.2 XP_006247885.1 1 - 2 101 86

View Full Product Details