Assay Design



Gene Transcripts

Gene Symbol
Sox15
Entrez Gene ID
363632
Gene Name
SRY (sex determining region Y)-box 15
Gene Aliases
-
Chromosome Location
Chr.10: 56260514 - 56263503 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108830.1 NP_001102300.1 1 - 2 290 63

View Full Product Details