Assay Design



Gene Transcripts

Gene Symbol
Narfl
Entrez Gene ID
360496
Gene Name
nuclear prelamin A recognition factor-like
Gene Aliases
-
Chromosome Location
Chr.10: 15143732 - 15152706 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013183.1 NP_001013201.1 9 - 10 1033 67
GenBank mRNA BC091240.1 - 9 - 10 1033 67

View Full Product Details