Assay Design



Gene Transcripts

Gene Symbol
Myh8
Entrez Gene ID
252942
Gene Name
myosin, heavy chain 8, skeletal muscle, perinatal
Gene Aliases
-
Chromosome Location
Chr.10: 53818818 - 53848490 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100485.1 NP_001093955.1 - 2762 93

View Full Product Details