Assay Design



Gene Transcripts

Gene Symbol
Smyd4
Entrez Gene ID
287525
Gene Name
SET and MYND domain containing 4
Gene Aliases
-
Chromosome Location
Chr.10: 62191669 - 62237959 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105810.1 NP_001099280.1 10 - 11 2526 64
XM_006246674.2 XP_006246736.1 10 - 11 2430 64
XM_008767962.1 XP_008766184.1 9 - 10 2655 64

View Full Product Details