Assay Design



Gene Transcripts

Gene Symbol
Emilin2
Entrez Gene ID
316736
Gene Name
elastin microfibril interfacer 2
Gene Aliases
-
Chromosome Location
Chr.9: 119594049 - 119653676 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008767441.1 XP_008765663.1 2 - 3 685 78

View Full Product Details