Assay Design
Gene Transcripts
- Gene Symbol
- Il2rg
- Entrez Gene ID
-
140924
- Gene Name
- interleukin 2 receptor, gamma
- Gene Aliases
-
Ab2-183, Cd132
- Chromosome Location
-
Chr.X: 71165378 - 71169038 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY325195.1 |
- |
- |
1292 |
90 |
- Gene Symbol
- CXHXorf65
- Entrez Gene ID
-
103690850
- Gene Name
- chromosome X open reading frame, human CXorf65
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 71162006 - 71165281 on Build Rnor_6.0
View Full Product Details