Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012044.1 | NP_001012044.1 | 12 - 13 | 1413 | 75 |
XM_006252318.2 | XP_006252380.1 | 12 - 13 | 1588 | 75 | |
XM_006252319.2 | XP_006252381.1 | 14 - 15 | 1787 | 75 | |
XM_006252320.2 | XP_006252382.1 | 14 - 15 | 1835 | 75 | |
XM_006252322.2 | XP_006252384.1 | 12 - 13 | 2994 | 75 | |
GenBank mRNA | BC083855.1 | - | 12 - 13 | 1413 | 75 |