Assay Design



Gene Transcripts

Gene Symbol
Pebp4
Entrez Gene ID
691997
Gene Name
phosphatidylethanolamine binding protein 4
Gene Aliases
-
Chromosome Location
Chr.15: 51645223 - 51740286 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008770836.1 XP_008769058.1 2 - 3 118 84

View Full Product Details