Assay Design



Gene Transcripts

Gene Symbol
Ppp6c
Entrez Gene ID
171121
Gene Name
protein phosphatase 6, catalytic subunit
Gene Aliases
-
Chromosome Location
Chr.3: 23480258 - 23510959 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133589.2 NP_598273.2 1 - 2 175 67
GenBank mRNA BC078747.1 - 1 - 2 175 67
X77236.1 - 1 - 2 140 67

View Full Product Details