Assay Design



Gene Transcripts

Gene Symbol
Bhlhe23
Entrez Gene ID
499952
Gene Name
basic helix-loop-helix family, member e23
Gene Aliases
Bhlhb4, RGD1559760
Chromosome Location
Chr.3: 176279997 - 176282211 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109211.1 NP_001102681.1 1 - 1 2035 99

View Full Product Details