Assay Design



Gene Transcripts

Gene Symbol
Ythdf1
Entrez Gene ID
296467
Gene Name
YTH N(6)-methyladenosine RNA binding protein 1
Gene Aliases
-
Chromosome Location
Chr.3: 176415911 - 176431423 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024756.1 NP_001019927.1 2 - 3 189 79
GenBank mRNA BC097360.1 - 2 - 3 189 79

View Full Product Details