Assay Design



Gene Transcripts

Gene Symbol
Srms
Entrez Gene ID
296472
Gene Name
src-related kinase lacking C-terminal regulatory tyrosine and N-terminal myristylation sites
Gene Aliases
-
Chromosome Location
Chr.3: 176709742 - 176716146 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011961.1 NP_001011961.1 2 - 3 624 76
GenBank mRNA BC090006.1 - 2 - 3 624 76

View Full Product Details