Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108397.1 | NP_001101867.1 | 6 - 7 | 2820 | 62 |
XM_008771124.1 | XP_008769346.1 | 5 - 6 | 2407 | 62 | |
XM_008771125.1 | XP_008769347.1 | 6 - 7 | 2447 | 62 | |
XM_008771126.1 | XP_008769348.1 | 6 - 7 | 2582 | 62 | |
XM_008771127.1 | XP_008769349.1 | 5 - 6 | 2404 | 62 | |
XM_008771128.1 | XP_008769350.1 | 6 - 7 | 2482 | 62 |