Assay Design



Gene Transcripts

Gene Symbol
Pde12
Entrez Gene ID
306231
Gene Name
phosphodiesterase 12
Gene Aliases
LRRGT00074, RGD1310975
Chromosome Location
Chr.16: 2357028 - 2367069 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013998.2 NP_001014020.2 1 - 2 1207 118
GenBank mRNA AY387060.1 - 1 - 2 1207 118
BC079396.1 - 1 - 2 1391 118

View Full Product Details