Assay Design



Gene Transcripts

Gene Symbol
Cyp2r1
Entrez Gene ID
361631
Gene Name
cytochrome P450, family 2, subfamily r, polypeptide 1
Gene Aliases
-
Chromosome Location
Chr.1: 184060522 - 184106599 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006230081.2 XP_006230143.1 1 - 2 611 76
XM_006230082.2 XP_006230144.1 1 - 2 611 76
XM_008759769.1 XP_008757991.1 1 - 2 611 76

View Full Product Details