Assay Design



Gene Transcripts

Gene Symbol
Kat8
Entrez Gene ID
310194
Gene Name
K (lysine) acetyltransferase 8
Gene Aliases
Myst1
Chromosome Location
Chr.1: 199360645 - 199372925 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017378.1 NP_001017378.1 5 - 6 691 117
GenBank mRNA BC083891.1 - 5 - 6 691 117

View Full Product Details