Assay Design



Gene Transcripts

Gene Symbol
Hepacam2
Entrez Gene ID
296846
Gene Name
HEPACAM family member 2
Gene Aliases
RGD1305697
Chromosome Location
Chr.4: 28403050 - 28437746 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106580.2 NP_001100050.1 5 - 6 1162 110
XM_006236029.2 XP_006236091.1 5 - 6 1215 110
GenBank mRNA BC168741.1 - 5 - 6 1162 110

View Full Product Details