Assay Design



Gene Transcripts

Gene Symbol
Gtpbp10
Entrez Gene ID
312054
Gene Name
GTP-binding protein 10 (putative)
Gene Aliases
Cldn12
Chromosome Location
Chr.4: 25635765 - 25653283 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100815.1 NP_001094285.1 8 - 9 856 152
GenBank mRNA CV103632.1 - 6 - 7 522 152

View Full Product Details