Assay Design



Gene Transcripts

Gene Symbol
Il21
Entrez Gene ID
365769
Gene Name
interleukin 21
Gene Aliases
IL-21
Chromosome Location
Chr.2: 123965021 - 123972356 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108943.2 NP_001102413.1 1 - 2 151 89
GenBank mRNA DQ387062.1 - 1 - 2 151 89

View Full Product Details