Assay Design



Gene Transcripts

Gene Symbol
Mmp19
Entrez Gene ID
304608
Gene Name
matrix metallopeptidase 19
Gene Aliases
-
Chromosome Location
Chr.7: 3216392 - 3224202 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107159.1 NP_001100629.1 4 - 5 612 65
XM_006240758.2 XP_006240820.1 4 - 5 532 65
GenBank mRNA FJ799755.1 - 4 - 5 612 65

View Full Product Details