Assay Design



Gene Transcripts

Gene Symbol
Kpna7
Entrez Gene ID
690504
Gene Name
karyopherin alpha 7
Gene Aliases
-
Chromosome Location
Chr.12: 11347643 - 11384349 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001074637.3 XP_001074637.3 5 - 6 624 66

View Full Product Details