Assay Design



Gene Transcripts

Gene Symbol
Rasl11a
Entrez Gene ID
304268
Gene Name
RAS-like family 11 member A
Gene Aliases
-
Chromosome Location
Chr.12: 9986043 - 9990284 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002829.1 NP_001002829.1 5 - 6 348 86
XM_006248834.2 XP_006248896.1 3 - 4 811 86

View Full Product Details