Assay Design



Gene Transcripts

Gene Symbol
Sgcz
Entrez Gene ID
364605
Gene Name
sarcoglycan zeta
Gene Aliases
-
Chromosome Location
Chr.16: 57136433 - 58207615 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108875.2 NP_001102345.2 2 - 3 448 110
XM_008771302.1 XP_008769524.1 2 - 3 282 110
GenBank mRNA BG664629.1 - - 448 110

View Full Product Details