Assay Design



Gene Transcripts

Gene Symbol
Sgcz
Entrez Gene ID
364605
Gene Name
sarcoglycan zeta
Gene Aliases
-
Chromosome Location
Chr.16: 57136433 - 58207615 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108875.2 NP_001102345.2 7 - 8 957 66
XM_008771302.1 XP_008769524.1 7 - 8 791 66

View Full Product Details