Assay Design



Gene Transcripts

Gene Symbol
Lig4
Entrez Gene ID
290907
Gene Name
ligase IV, DNA, ATP-dependent
Gene Aliases
-
Chromosome Location
Chr.16: 85331766 - 85339496 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006253477.2 XP_006253539.1 1 - 2 222 67

View Full Product Details