Assay Design



Gene Transcripts

Gene Symbol
Angpt2
Entrez Gene ID
89805
Gene Name
angiopoietin 2
Gene Aliases
Agpt2, Ang-2
Chromosome Location
Chr.16: 75966480 - 76016147 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134454.1 NP_604449.1 5 - 6 1001 58
GenBank mRNA BC161931.1 - 5 - 6 1001 58

View Full Product Details