Assay Design



Gene Transcripts

Gene Symbol
Rftn1
Entrez Gene ID
501095
Gene Name
raftlin lipid raft linker 1
Gene Aliases
2310015n21rik, RGD1310777, RGD1563347
Chromosome Location
Chr.9: 12740885 - 12942358 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135011.1 NP_001128483.1 1 - 2 146 103

View Full Product Details