Assay Design



Gene Transcripts

Gene Symbol
Mrps10
Entrez Gene ID
363187
Gene Name
mitochondrial ribosomal protein S10
Gene Aliases
Ac1179
Chromosome Location
Chr.9: 15637427 - 15651154 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006244464.2 XP_006244526.1 2 - 3 171 86

View Full Product Details