Assay Design



Gene Transcripts

Gene Symbol
Fbxw17
Entrez Gene ID
361219
Gene Name
F-box and WD-40 domain protein 17
Gene Aliases
RGD1566133
Chromosome Location
Chr.17: 15076773 - 15094620 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082409.1 NP_001075878.1 5 - 6 638 88
XM_008771539.1 XP_008769761.1 5 - 6 683 88
XM_008771540.1 XP_008769762.1 5 - 6 683 88
XM_008771541.1 XP_008769763.1 5 - 6 683 88
XM_008771542.1 XP_008769764.1 5 - 6 1645 88
GenBank mRNA BC167744.1 - 5 - 6 626 88

View Full Product Details