Assay Design



Gene Transcripts

Gene Symbol
Fbxw17
Entrez Gene ID
361219
Gene Name
F-box and WD-40 domain protein 17
Gene Aliases
RGD1566133
Chromosome Location
Chr.17: 15076773 - 15094620 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008771540.1 XP_008769762.1 7 - 8 1057 74
GenBank mRNA BC167744.1 - 7 - 8 1000 74

View Full Product Details