Assay Design
Gene Transcripts
- Gene Symbol
- Gcnt2
- Entrez Gene ID
-
306860
- Gene Name
- glucosaminyl (N-acetyl) transferase 2, I-branching enzyme
- Gene Aliases
-
Ignt
- Chromosome Location
-
Chr.17: 21634546 - 21677477 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY435152.1 |
- |
- |
920 |
69 |
View Full Product Details