Assay Design



Gene Transcripts

Gene Symbol
Arrdc3
Entrez Gene ID
309945
Gene Name
arrestin domain containing 3
Gene Aliases
LRRGT00048
Chromosome Location
Chr.2: 8732910 - 8745448 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007797.1 NP_001007798.1 1 - 2 279 125
GenBank mRNA AY383703.1 - 1 - 2 279 125

View Full Product Details